Learn More
Medchemexpress LLC Bupivacaine (hydrochloride) | 18010-40-7 | 99.9% | 25 MG
SDP
Catalog No. 5000450544
Encompass_Preferred
Bupivacaine hydrochloride is an NMDA receptor inhibitor that can also block sodium, L-calcium, and potassium channels. It potently blocks SCN5A channels with an IC50 of 69.5 μM. This compound is suitable for research related to chronic pain.
  • NMDA receptor inhibitor.
  • Blocks sodium, L-calcium, and potassium channels.
  • Potent blocker of SCN5A channels.
  • Useful for chronic pain research.
Catalog No. 50-004-50544 Supplier Medchemexpress LLC Supplier No. 25MGHYB0405AR
Bupivacaine hydrochloride is an NMDA receptor inhibitor that can also block sodium, L-calcium, and potassium channels. It potently blocks SCN5A channels with an IC50 of 69.5 μM. This compound is suitable for research related to chronic pain.
  • NMDA receptor inhibitor.
  • Blocks sodium, L-calcium, and potassium channels.
  • Potent blocker of SCN5A channels.
  • Useful for chronic pain research.