Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bupivacaine hydrochloride | 18010-40-7 | MFCD00078956 | 100.0% | 324.89 | C18H29ClN2O | 10 MG
SDP

Catalog No. 5000298910
Encompass_Preferred

Bupivacaine hydrochloride is an analytical reference standard of the local anesthetic bupivacaine supplied for research and analytical applications. It is intended for use in chromatographic assays, method development, and studies of sodium-channel and NMDA-receptor pharmacology.

  • Analytical reference standard for chromatography and assay.
  • High purity: 99.99% (as stated by manufacturer).
  • CAS number: 18010-40-7.
  • Molecular formula: C18H29ClN2O.
  • Molecular weight: 324.89.
  • Packaged in a brown glass bottle and supplied in small-mass quantities suitable for analytical use.

Catalog No. 50-002-98910 Supplier Medchemexpress LLC Supplier No. HYB0405AR10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bupivacaine hydrochloride is an analytical reference standard of the local anesthetic bupivacaine supplied for research and analytical applications. It is intended for use in chromatographic assays, method development, and studies of sodium-channel and NMDA-receptor pharmacology.

  • Analytical reference standard for chromatography and assay.
  • High purity: 99.99% (as stated by manufacturer).
  • CAS number: 18010-40-7.
  • Molecular formula: C18H29ClN2O.
  • Molecular weight: 324.89.
  • Packaged in a brown glass bottle and supplied in small-mass quantities suitable for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.