Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupivacaine hydrochloride monohydrate, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111018747 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
15 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8747 5 g
11-101-9313 1 g
11-101-8210 15 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8747 Supplier MilliporeSigma Supelco Supplier No. B52745G
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 73360-54-0
Physical Form Neat
Linear Formula C18H28N2 O · HCl · H2O
MDL Number MFCD00941462
Synonym 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide
Type Analytical Standard
Industry Type Drug Analysis
Quantity 5 g
Formula Weight 342.90
Percent Purity ≥99%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.