Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupropion, 99.91%, MP Biomedicals™

Catalog No. ICN15376015 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
15 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15376015 15 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15376015 Supplier MP Biomedicals, Inc Supplier No. 0215376015

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31677-93-7
Molecular Formula C13H19Cl2NO
Molecular Weight (g/mol) 276.201
InChI Key HEYVINCGKDONRU-UHFFFAOYSA-N
Synonym bupropion hydrochloride, wellbutrin, 2-tert-butylamino-1-3-chlorophenyl propan-1-one hydrochloride, bupropion hcl, zyban, wellbutrin xl, bupropion hydrocloride, wellbutrin sr, budeprion, forfivo xl
PubChem CID 62884
IUPAC Name 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one;hydrochloride
SMILES CC(C(=O)C1=CC(=CC=C1)Cl)NC(C)(C)C.Cl

Specifications

Color White
Assay Percent Range 0.9991
Quantity 15 mg
Formula Weight 276.2
Physical Form Powder
Chemical Name or Material Bupropion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.