Learn More
Enzo Life Sciences Bupropion . HCl (250mg). CAS: 31677-93-7
SDP
Supplier:  Enzo Life Sciences BMLNS1110250
Encompass_Preferred
Inhibits dopamine and norepinephrine transporters (Ki=2.8 and 1.4µM respectively). Inhibits the serotonin transporter at higher concentration (Ki=45µM). In clinical use as an antidepressant and for smoking cessation. Also acts as a nicotinic receptor antagonist. Purity: ≥98% (TLC). Solubility: Soluble in water (>25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1208