Learn More
Apexbio Technology LLC Bupropion hydrochloride 31677-93-7 50mg
SDP
Catalog No. 502240429
Encompass_Preferred
Bupropion hydrochloride (CAS 31677-93-7) is a small-molecule aminoketone that acts as a dual inhibitor of norepinephrine and dopamine reuptake with IC values of 6 5 0 6 M and 3 4 0 4 M respectively It is widely utilized in research for its established roles in modulating monoaminergic neurotransmission associated with depression and smoking cessation In cellular studies bupropion hydrochloride has been shown to decrease cell viability in SH-SY5Y neuroblastoma cells selectively induce endoplasmic reticulum stress and activate JNK pathways leading to caspase 3-dependent apoptosis Additionally it has been reported to downregulate tyrosine hydroxylase expression impacting dopamine biosynthesis
This item is not returnable. View return policy
Catalog No. 50-224-0429 Supplier Apexbio Technology LLC Supplier No. B332650
Bupropion hydrochloride (CAS 31677-93-7) is a small-molecule aminoketone that acts as a dual inhibitor of norepinephrine and dopamine reuptake with IC values of 6 5 0 6 M and 3 4 0 4 M respectively It is widely utilized in research for its established roles in modulating monoaminergic neurotransmission associated with depression and smoking cessation In cellular studies bupropion hydrochloride has been shown to decrease cell viability in SH-SY5Y neuroblastoma cells selectively induce endoplasmic reticulum stress and activate JNK pathways leading to caspase 3-dependent apoptosis Additionally it has been reported to downregulate tyrosine hydroxylase expression impacting dopamine biosynthesis