Learn More
Abcam Bupropion hydrochloride, Non-selective DAT/NET inhibitor, 50MG
Catalog No. 502707513
Encompass_Preferred
MW 276.2 g/mol, Purity >98%. Non-selective dopamine and norepinephrine transporter (DAT/NET) inhibitor (Ki values are 1.4, 2.8 and 45 μM for NET, DAT and SERT transporters, respectively). Nicotinic acetylcholine receptor (nAChR) antagonist (IC₅₀ = 1.8 μM at α₂β₂ receptor). Displays antidepressant activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7513 Supplier Abcam Supplier No. AB12053450MG
MW 276.2 g/mol, Purity >98%. Non-selective dopamine and norepinephrine transporter (DAT/NET) inhibitor (Ki values are 1.4, 2.8 and 45 μM for NET, DAT and SERT transporters, respectively). Nicotinic acetylcholine receptor (nAChR) antagonist (IC₅₀ = 1.8 μM at α₂β₂ receptor). Displays antidepressant activity.
The product is subject to the following: Abcam Restricted Use Statement