Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bupropion Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18601138 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-138 1 kg Amber Glass Bottle
18-601-134 1 g Amber Glass Bottle
18-601-135 5 g Amber Glass Bottle
18-601-136 25 g Amber Glass Bottle
18-601-137 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-138 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B12371KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

B1237, 31677-93-7, C13H18ClNO·HCl

Spectrum™ Chemical Bupropion Hydrochloride, USP is an antidepressant medication. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 31677-93-7
Molecular Formula C13H19Cl2NO
Molecular Weight (g/mol) 276.20
InChI Key HEYVINCGKDONRU-UHFFFAOYNA-N
IUPAC Name hydrogen 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one chloride
SMILES [H+].[Cl-].CC(NC(C)(C)C)C(=O)C1=CC=CC(Cl)=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98-102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Bupropion Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.