Learn More
Medchemexpress LLC Bupropion morpholinol | 357399-43-0 | 99.7% | 10 MG
SDP
Catalog No. 5000339749
Encompass_Preferred
Bupropion morpholinol, also known as Hydroxy Bupropion, is a major metabolite of Bupropion. It functions by inhibiting Dopamine and Norepinephrine transporters, as well as the α4β2 nicotinic receptor in vitro. This compound plays a role in antidepressant and smoking cessation activities. It is produced from Bupropion in vivo by the genetically polymorphic enzyme CYP2B6. It inhibits [3H]norepinephrine uptake with an IC50 of 1.7 μM. This product is intended for research use only and is not sold to patients.
  • Major metabolite of bupropion
  • Inhibits dopamine and norepinephrine transporters
  • Inhibits the α4β2 nicotinic receptor in vitro
  • Contributes to antidepressant and smoking cessation activities
  • Produced from bupropion by the genetically polymorphic enzyme CYP2B6 in vivo
  • Inhibits [3H]norepinephrine uptake with an IC50 of 1.7 μM
  • Intended for research use only
Catalog No. 50-003-39749 Supplier Medchemexpress LLC Supplier No. HY12766S10MG
Bupropion morpholinol, also known as Hydroxy Bupropion, is a major metabolite of Bupropion. It functions by inhibiting Dopamine and Norepinephrine transporters, as well as the α4β2 nicotinic receptor in vitro. This compound plays a role in antidepressant and smoking cessation activities. It is produced from Bupropion in vivo by the genetically polymorphic enzyme CYP2B6. It inhibits [3H]norepinephrine uptake with an IC50 of 1.7 μM. This product is intended for research use only and is not sold to patients.
  • Major metabolite of bupropion
  • Inhibits dopamine and norepinephrine transporters
  • Inhibits the α4β2 nicotinic receptor in vitro
  • Contributes to antidepressant and smoking cessation activities
  • Produced from bupropion by the genetically polymorphic enzyme CYP2B6 in vivo
  • Inhibits [3H]norepinephrine uptake with an IC50 of 1.7 μM
  • Intended for research use only