Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bupropion morpholinol-d6 | 1216893-18-3 | MFCD08063409 | 99.3% | 261.78 | C13H12D6ClNO2 | 5 MG
SDP

Catalog No. 5000264860
Encompass_Preferred

Bupropion morpholinol-d6 is the deuterium-labeled (D6) analog of the major bupropion metabolite hydroxybupropion. It is supplied as a high-purity, isotope-labeled reference material intended for bioanalytical method development and quantitative mass spectrometry applications. The material is provided as a small, weighed solid for analytical use and is intended for research purposes only.

  • Isotope-labeled internal standard for LC-MS quantification.
  • Chemical purity 99.3%.
  • Deuterium-labeled formula C13H12D6ClNO2 and molecular weight 261.78 g/mol.
  • Provided as a 5 mg solid suitable for small-scale analytical workflows.
  • Intended for research use only; not for human or diagnostic use.

Catalog No. 50-002-64860 Supplier Medchemexpress LLC Supplier No. HY12766S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bupropion morpholinol-d6 is the deuterium-labeled (D6) analog of the major bupropion metabolite hydroxybupropion. It is supplied as a high-purity, isotope-labeled reference material intended for bioanalytical method development and quantitative mass spectrometry applications. The material is provided as a small, weighed solid for analytical use and is intended for research purposes only.

  • Isotope-labeled internal standard for LC-MS quantification.
  • Chemical purity 99.3%.
  • Deuterium-labeled formula C13H12D6ClNO2 and molecular weight 261.78 g/mol.
  • Provided as a 5 mg solid suitable for small-scale analytical workflows.
  • Intended for research use only; not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.