Learn More
Med Vet International Miltex Friction Grip Carbide Bur, Plain Fissure, Taper, 10 Pack, Dfg169L
Catalog No. 502831054
Encompass_Preferred
High quality tungsten carbide tips are brazed into the neck of the stainless steel shank. This maximizes the strength of the burs and reduces the risk of breaking. This ensures an even tooth preparation and more efficient cutting. Incredibly long-lasting and durable. Plain fissure. Taper. Long handle. 10 Pack. 169L.
Catalog No. 50-283-1054 Supplier Med Vet International Supplier No. MILDFG169L
High quality tungsten carbide tips are brazed into the neck of the stainless steel shank. This maximizes the strength of the burs and reduces the risk of breaking. This ensures an even tooth preparation and more efficient cutting. Incredibly long-lasting and durable. Plain fissure. Taper. Long handle. 10 Pack. 169L.