Learn More
Med Vet International Miltex Friction Grip Carbide Bur, 0.6mm, #1, 10 Pack, Dfg1
Catalog No. 502831260
Encompass_Preferred
High quality tungsten carbide tips are brazed into the neck of the stainless steel shank. This maximizes the strength of the burs and reduces the risk of breaking. This ensures an even tooth preparation and more efficient cutting. Incredibly long-lasting and durable. #1/2. 0.6mm. 100 Pack.
Catalog No. 50-283-1260 Supplier Med Vet International Supplier No. MILDFG1
High quality tungsten carbide tips are brazed into the neck of the stainless steel shank. This maximizes the strength of the burs and reduces the risk of breaking. This ensures an even tooth preparation and more efficient cutting. Incredibly long-lasting and durable. #1/2. 0.6mm. 100 Pack.