Learn More
Med Vet International Miltex Friction Grip Carbide Bur, Round Tip, #2, 10/Pack, Dfg2
Catalog No. 502829664
Encompass_Preferred
High quality tungsten carbide tips are brazed into the neck of the stainless steel shank. This maximizes the strength of the burs and reduces the risk of breaking. This ensures an even tooth preparation and more efficient cutting. Incredibly long-lasting and durable. Round tip. 10 Pack. 2.
Catalog No. 50-282-9664 Supplier Med Vet International Supplier No. MILDFG2
High quality tungsten carbide tips are brazed into the neck of the stainless steel shank. This maximizes the strength of the burs and reduces the risk of breaking. This ensures an even tooth preparation and more efficient cutting. Incredibly long-lasting and durable. Round tip. 10 Pack. 2.