Learn More
Med Vet International Miltex Friction Grip Tungsten Carbide Bur, Crosscut Fissure, Straight, Long, #57L, 10 Pack, Dfg57L
Catalog No. 502831602
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Crosscut fissure taper. Straight bur. Long handle. 10 pack. #57L.
Catalog No. 50-283-1602 Supplier Med Vet International Supplier No. MILDFG57L
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Crosscut fissure taper. Straight bur. Long handle. 10 pack. #57L.