Learn More
Med Vet International Miltex Friction Grip Tungsten Carbide Bur, Straight, #700, 10 Pack, Dfg700
Catalog No. 502829827
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Straight bur. 10 pack. #700
Catalog No. 50-282-9827 Supplier Med Vet International Supplier No. MILDFG700
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Straight bur. 10 pack. #700