Learn More
Med Vet International Miltex Friction Grip Tungsten Carbide Bur, Crosscut Fissure Taper, Surgical, #701Su, 10 Pack, Dfg701Su
Catalog No. 502831759
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Crosscut fissure taper. Long handle. 10 pack. Surgical variant. 701SU.
Catalog No. 50-283-1759 Supplier Med Vet International Supplier No. MILDFG701SU
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Crosscut fissure taper. Long handle. 10 pack. Surgical variant. 701SU.