Learn More
Med Vet International Miltex Friction Grip Tungsten Carbide Bur, Crosscut Fissure Taper, #700L, 10 Pack, Dfg700L
Catalog No. 502829737
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Crosscut fissure taper. 10 pack. 700L.
Catalog No. 50-282-9737 Supplier Med Vet International Supplier No. MILDFG700L
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Crosscut fissure taper. 10 pack. 700L.