Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Christmas Tree, Extra Fine Grit, Conical, German, German, 853/016
Catalog No. 502832034
Encompass_Preferred
Round friction grip diamond burs are rotary instruments attached to a handpiece used for opening and shaping cavity preparations, plus they can be used to depth cuts. The burs feature multiple, evenly distributed layers for optimum concentricity and reliable cutting. Christmas tree design.
Catalog No. 50-283-2034 Supplier Med Vet International Supplier No. MIL853/016
Round friction grip diamond burs are rotary instruments attached to a handpiece used for opening and shaping cavity preparations, plus they can be used to depth cuts. The burs feature multiple, evenly distributed layers for optimum concentricity and reliable cutting. Christmas tree design.