Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Flame, Long Ogival, (012), Med, Each, 863/012
Catalog No. 502643505
Encompass_Preferred
A safe end flame shaped diamond can be used for sub-gingival curettage and can remove sub-gingival calculus for initial root planning. Extra fine grit allows for more delicate procedures to be done. 1.0mm. #12. Long ogival
Catalog No. 50-264-3505 Supplier Med Vet International Supplier No. MIL863/012
A safe end flame shaped diamond can be used for sub-gingival curettage and can remove sub-gingival calculus for initial root planning. Extra fine grit allows for more delicate procedures to be done. 1.0mm. #12. Long ogival