Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Disc, Diamond, Single Side, Diamond Coat, Xflex, 22mm, German, Each, 914/220
Catalog No. 502832324
Encompass_Preferred
Round friction grip diamond burs are rotary instruments attached to a handpiece used for opening and shaping cavity preparations, plus they can be used to depth cuts. The burs feature multiple, evenly distributed layers for optimum concentricity and reliable cutting. Disc shaped bur. The disc design is primarily used for contouring, fabricating, finishing, and the shaping of dental restorative materials.
Catalog No. 50-283-2324 Supplier Med Vet International Supplier No. MIL914/220
Round friction grip diamond burs are rotary instruments attached to a handpiece used for opening and shaping cavity preparations, plus they can be used to depth cuts. The burs feature multiple, evenly distributed layers for optimum concentricity and reliable cutting. Disc shaped bur. The disc design is primarily used for contouring, fabricating, finishing, and the shaping of dental restorative materials.