Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Round, 1.86mm, (018), Med, Each, 801/018
Catalog No. 502830673
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Round tip. Med grit. 1.86mm #18.
Catalog No. 50-283-0673 Supplier Med Vet International Supplier No. MIL801/018
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Round tip. Med grit. 1.86mm #18.