Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Wheel, (035), Medium Grit, Each, 909/035
Catalog No. 502832060
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Wheel bur. Medium grit. #35.
Catalog No. 50-283-2060 Supplier Med Vet International Supplier No. MIL909/035
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Wheel bur. Medium grit. #35.