Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Round, Conical, (010), Med, Each, 805/010
Catalog No. 502830674
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Round tip. Conical. Med grit. #10.
Catalog No. 50-283-0674 Supplier Med Vet International Supplier No. MIL805/010
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Round tip. Conical. Med grit. #10.