Learn More
Med Vet International Miltex Friction Grip Diamond Bur, Conical, Conical, Standard, (016), Med, Each, 807/016
Catalog No. 502830675
Encompass_Preferred
Produced using a coating technique, which assure approximately 85% of the working surface is coated with a uniform diamond grit. The extremely strong stainless shaft resists bending and breakage. Medium grit. #16
Catalog No. 50-283-0675 Supplier Med Vet International Supplier No. MIL807/016
Produced using a coating technique, which assure approximately 85% of the working surface is coated with a uniform diamond grit. The extremely strong stainless shaft resists bending and breakage. Medium grit. #16