Learn More
Med Vet International Miltex Friction Grip Tungsten Carbide Bur, inverted Cone, 10 Pack, Dfg36
Catalog No. 502831605
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Inverted cone. 10 pack. #36.
Catalog No. 50-283-1605 Supplier Med Vet International Supplier No. MILDFG36
 Restricted Item
Add to Cart
Add to Cart
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Inverted cone. 10 pack. #36.