Learn More
Med Vet International Miltex Friction Grip Tungsten Carbide Bur, Pear, #332, 10 Pack, Dfg332
Catalog No. 502831604
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Pear shaped tip. 10 pack. #332
Catalog No. 50-283-1604 Supplier Med Vet International Supplier No. MILDFG332
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Pear shaped tip. 10 pack. #332