Learn More
Med Vet International Miltex Friction Grip Tungsten Carbide Bur, Round, Surgical, #6Su, 10 Pack, Dfg6Su
Catalog No. 502829578
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Round tip. 10 pack. Surgical variant. #6SU
Catalog No. 50-282-9578 Supplier Med Vet International Supplier No. MILDFG6SU
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Round tip. 10 pack. Surgical variant. #6SU