Learn More
Med Vet International Miltex Tungsten Carbide Bur, Round Tip, Right Angle Latch, #3, 10 Pack, Dra3
Catalog No. 502829569
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Round tip. Right angle latch. 10 pack. #3
Catalog No. 50-282-9569 Supplier Med Vet International Supplier No. MILDRA3
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Round tip. Right angle latch. 10 pack. #3