Learn More
Med Vet International Miltex Tungsten Carbide Bur, Round Tip, Right Angle, #8, 10 Pack, Dra8
Catalog No. 502831797
Encompass_Preferred
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Round tip. Right angle latch. 10 pack. #8
Catalog No. 50-283-1797 Supplier Med Vet International Supplier No. MILDRA8
Miltex diamond burs come in a variety of shapes and grits for more flexibility for procedures. Diamond particles are uniform in coverage for optimal cutting performance. Tungsten Carbide inlays offer superior durability and efficiency. Round tip. Right angle latch. 10 pack. #8