Learn More
Hach Company Buret-Acrylic-25mL
Supplier:  Hach Company 1405940
Encompass
Durable crystal-clear acrylic body is individually calibrated to meet the accuracy requirements of ASTM E287 Leak-proof stopcock with self-lubricating PTFE TFE plug Excellent for general lab or field use in schools industrial municipal or food applications Unaffected by dilute mineral acids and bases (except hydrofluoric acid and ammonium hydroxide) Can contain liquids within a pH range of 1-14 and up to 5% wt/wt of acids or 30% bases can be used Not designed for exposure to alcohol or organic solvents
Catalog No. 50-002-57169