Learn More
Wilmad Labglass BURET AMMONIA DISSOCIATION
Supplier:  Wilmad Labglass LG4150100
Encompass
Buret, Ammonia Dissociation. Used to determine the degree of dissociation of ammonia during the nitriding of steel alloys. Buret is 100mL and reads the amount of ammonia absorbed. Graduated from 10 to 90 percent in 1% subdivisions.
Catalog No. 50-801-636
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.