Learn More
Wilmad Labglass BURET AUTO LAB GRADE 10ML
Catalog No. 50805937
Encompass
Buret, Laboratory Grade, Automatic, with Straight PTFE Stopcock, Complete. Buret Capacity: 10mL; Subdivision: 0.1mL; Reservoir Capacity: 16 ounces; Stopcock Size: 2mm. Buret is filled by pressure supplied by the rubber bulb.
Catalog No. 50-805-937 Supplier Wilmad Labglass Supplier No. LG4241T100
Buret, Laboratory Grade, Automatic, with Straight PTFE Stopcock, Complete. Buret Capacity: 10mL; Subdivision: 0.1mL; Reservoir Capacity: 16 ounces; Stopcock Size: 2mm. Buret is filled by pressure supplied by the rubber bulb.