Learn More
Cole-Parmer BURET AUTOMATIC BOROSILCTE 25M
Supplier:  Cole-Parmer 0604448
Encompass_Preferred
Precise dosing using press button dispensing. Constructed with a borosilicate glass 3.3 burette (buret). Flexible left- or right-handed operation possible. Accurately titrate with the micro-screw and discharge jet. Designed with a 25 mL capacity.
Catalog No. NC1849755