Learn More
Hach Company Buret, Class A, Glass, 50 mL
Supplier:  Hach Company 2636541
Encompass
Glass, funnel-fill buret. Certified Class A (ASTM Specification E287) with permanent serial number and certificate for NIST traceability. PTFE stopcock does not require grease. Dust cap included. 10 mL buret meets requirements for potentiometric titrations (ASTM D664).
Catalog No. 50-199-6426