Learn More
Chemglass Life Sciences Buret, Class A, 25ml, TFE Stopcock
SDP
Catalog No. NC0961242
Encompass_Preferred
Buret, Class A, 25ml, TFE Stopcock. The capacity tolerance on these burets is established by ASTM E-287 and they are calibrated in accordance with ASTM E-542. Tips are carefully drawn from accurate bore tubing to insure proper drainage rates. This buret has colored markings, fine, sharp lines and large easy-to-read numbers. The stopcock barrel has a smooth micro-finish to assure leak-resistant performance. Each buret is supplied with a dust cover. furnished with a 2mm bore stopcock plug.
Catalog No. NC0961242 Supplier Chemglass Life Sciences Supplier No. CG815425
Buret, Class A, 25ml, TFE Stopcock. The capacity tolerance on these burets is established by ASTM E-287 and they are calibrated in accordance with ASTM E-542. Tips are carefully drawn from accurate bore tubing to insure proper drainage rates. This buret has colored markings, fine, sharp lines and large easy-to-read numbers. The stopcock barrel has a smooth micro-finish to assure leak-resistant performance. Each buret is supplied with a dust cover. furnished with a 2mm bore stopcock plug.