Learn More
Wilmad Labglass Buret, with Straight Bore PTFE Stopcock, 10 mL
Catalog No. 50806287
Encompass
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 10mL, Subdivision: 0.5mL; Tolerance:0.02mL, Stopcock Size: 2mm. With funnel shaped top so that a rubber stopper carrying a short length of glass tubing can be inserted.
Catalog No. 50-806-287 Supplier Wilmad Labglass Supplier No. LG4011T102
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 10mL, Subdivision: 0.5mL; Tolerance:0.02mL, Stopcock Size: 2mm. With funnel shaped top so that a rubber stopper carrying a short length of glass tubing can be inserted.