Learn More
Wilmad Labglass BURET PB TFE S/C MICRO 5ML
Catalog No. 50806286
Encompass
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 5mL, Subdivision: 0.01mL; Tolerance:0.01mL, Stopcock Size: 2mm. With funnel shaped top so that a rubber stopper carrying a short length of glass tubing can be inserted.
Catalog No. 50-806-286 Supplier Wilmad Labglass Supplier No. LG4011T100
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 5mL, Subdivision: 0.01mL; Tolerance:0.01mL, Stopcock Size: 2mm. With funnel shaped top so that a rubber stopper carrying a short length of glass tubing can be inserted.