Learn More
Wilmad Labglass BURET PREC BORE CASSL A 25ML
Catalog No. 50806855
Encompass
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 25mL; Subdivision: 0.10mL; Tolerance: 0.03ml; Stopcock Size: 2mm. Calibration is in compliance with ASTM E 694 specifications.
Catalog No. 50-806-855 Supplier Wilmad Labglass Supplier No. LG3956T102
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 25mL; Subdivision: 0.10mL; Tolerance: 0.03ml; Stopcock Size: 2mm. Calibration is in compliance with ASTM E 694 specifications.