Learn More
Wilmad Labglass BURET PREC BORE CLASS A 100ML
Supplier:  Wilmad Labglass LG3956T106
Encompass
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 100mL; Subdivision: 0.05mL; Tolerance: 0.10ml; Stopcock Size: 2mm. Calibration is in compliance with ASTM E 694 specifications.
Catalog No. 50-806-616
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.