Learn More
Wilmad Labglass BURET PREC BORE CLASS A 10ML
Catalog No. 50804245
Encompass
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 10mL; Subdivision: 0.05mL; Tolerance: 0.02ml; Stopcock Size: 2mm. Calibration is in compliance with ASTM E 694 specifications.
Catalog No. 50-804-245 Supplier Wilmad Labglass Supplier No. LG3956T100
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 10mL; Subdivision: 0.05mL; Tolerance: 0.02ml; Stopcock Size: 2mm. Calibration is in compliance with ASTM E 694 specifications.