Learn More
Wilmad Labglass BURET PREC BORE CLASS A 50ML
Catalog No. 50804008
Encompass
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 50mL; Subdivision: 0.10mL; Tolerance: 0.05ml; Stopcock Size: 2mm. Calibration is in compliance with ASTM E 694 specifications.
Catalog No. 50-804-008 Supplier Wilmad Labglass Supplier No. LG3956T104
Buret, Class A, with Straight Bore PTFE Stopcock. Capacity: 50mL; Subdivision: 0.10mL; Tolerance: 0.05ml; Stopcock Size: 2mm. Calibration is in compliance with ASTM E 694 specifications.