Learn More
Hach Company Glass T for Automatic Buret
Supplier:  Hach Company 50444
Encompass
Glass reservoir and buret with PTFE stopcock create an easy to use workstation for standard solutions. Subdivisions of 1.0 mL and 0.1 mL.
Catalog No. 50-199-4321