Learn More
Cole-Parmer Pyrex 2103-10 2103 Class A Glass Burette, 10 mL; 1/Cs
Supplier:  Cole-Parmer 3454321
Encompass_Preferred
Burettes (burets) are calibrated in accordance with ASTM E-542. Tips are carefully drawn from accurate bore tubing to ensure proper drainage rates. Burettes have colored markings, fine sharp lines, and large, easy-to-read numbers. The stopcock barrel has a smooth micro-finish to assure leak-resistant performance. Each burette is supplied with a dust cover, and all sizes are furnished with a 2-mm bore stopcock plug.
Catalog No. NC0208067