Learn More
Cole-Parmer Cole-Parmer Burette, 50 mL With Glass stopcock
Supplier:  Cole-Parmer 6310055
Encompass_Preferred
Burettes (burets) are manufactured from high quality boresilicate tubing to provide a high degree of accuracy, reliablity, and durability. Burettes have a glass stopcock and imprinted red enamel graduations.
Catalog No. NC0893794
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov