Learn More
Cole-Parmer Class B Burette w/PTFE stopcock; 50 mL, 1/ea
Supplier:  Cole-Parmer 6310038
Encompass_Preferred
Cole-Parmer Class B Burette w/PTFE stopcock; 50 mL, 1/ea, Class B Burette (Burets) meets your basic measuring and dispensing requirements!Designed with a 50 mL capacity, Manufactured from high quality borosilicate glass, burettes (Burets) are designed for accuracy and durability. Tips have been carefully drawn for reliable drainage rate. Burettes feature non-contaminating 2mm straight bore PTFE stopcock, and red colored permanent graduations.
Catalog No. NC1439598
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov