Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Burette Double Clamp with Coated Jaws
SCI_ED

Catalog No. S24618 Shop All GSC Products
Encompass
Change view
Click to view available options
Material:
Cast Iron
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
S24618 Cast Iron
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S24618 Supplier GSC Go Science Crazy Supplier No. 4BCDR865

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Securely holds two burettes simultaneously.

  • Arms of the clamp will rotate 360 degrees and can be locked in place
  • Cast iron construction
  • Rubber coated jaws to prevent slipping
  • Accept a tube range of 0.375 to 1.375 in.in. diameter

Specifications

Material Cast Iron
For Use With (Application) Holding burettes
Size (English) Grip 0.375 to 1.375 in.
Coating Rubber
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.