Learn More
METTLER TOLEDO 3P INDUSTRIAL BURETTE PLUG-N-PLAY UPGRADE
Catalog No. NC3336369
Encompass
  • Enhanced functionality with plug & play design.
  • Automatic recognition by titrator, promoting efficient operations.
  • Integrated RFID chip facilitates seamless interchangeability.
  • Minimizes risk of incorrect titrant or sensor use.
  • Rapid change of titrants improves workflow efficiency.
  • Supports high-precision burettes resistant to chemicals.
  • Helps maintain an error-free operational environment.
  • Ensures reliable dispensing and titration analyses.
  • Complies with ISO 8655-3 and GLP standards with certification.
Catalog No. NC3336369 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51109117
  • Enhanced functionality with plug & play design.
  • Automatic recognition by titrator, promoting efficient operations.
  • Integrated RFID chip facilitates seamless interchangeability.
  • Minimizes risk of incorrect titrant or sensor use.
  • Rapid change of titrants improves workflow efficiency.
  • Supports high-precision burettes resistant to chemicals.
  • Helps maintain an error-free operational environment.
  • Ensures reliable dispensing and titration analyses.
  • Complies with ISO 8655-3 and GLP standards with certification.