Learn More
Foxx Life Sciences Borosil Class A Burettes (Burets) with Ultimate Control Boroflo PTFE Screw Thread Keys, Batch Certificate, ASTM, 50mL, 5/CS
Catalog No. 501542823
Encompass_Preferred
Borosil Boroflo Burettes ("Burets") provide users with the ultimate flow control. Replaceable Boroflo PTFE Screw-Thread Keys create a chemically-resistant, leakproof system which requires no lubrication whatsoever. Each Borosil Burette is calibrated by automatic laser calibration. Certificates can be downloaded by visiting Borosil.com, or by downloading the Borosil app. Avail. from 10mL to 100mL. Burettes comply with ASTM E-287-2. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-154-2823 Supplier Foxx Life Sciences Supplier No. 2121U12A
Borosil Boroflo Burettes ("Burets") provide users with the ultimate flow control. Replaceable Boroflo PTFE Screw-Thread Keys create a chemically-resistant, leakproof system which requires no lubrication whatsoever. Each Borosil Burette is calibrated by automatic laser calibration. Certificates can be downloaded by visiting Borosil.com, or by downloading the Borosil app. Avail. from 10mL to 100mL. Burettes comply with ASTM E-287-2. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil Glassware is stress-free and incredibly resistant to thermal shock.