Learn More
Foxx Life Sciences Borosil® Class A Burettes (Burets) with Ultimate Control Boroflo PTFE Screw Thread Keys, Individual Certificate, ISO 385, 50mL, 5/CS
Catalog No. 501720545
Encompass_Preferred
Borosil® Boroflo Burettes ("Burets") provide users with the ultimate flow control. Replaceable Boroflo PTFE Screw-Thread Keys create a chemically-resistant, leakproof system which requires no lubrication whatsoever. Each Borosil® Burette is calibrated by automatic laser calibration. Certificates can be downloaded by visiting Borosil.com, or by downloading the Borosil app. Avail. from 10mL to 100mL. Burettes comply with ISO-385. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0545 Supplier Foxx Life Sciences Supplier No. 2121012D
Borosil® Boroflo Burettes ("Burets") provide users with the ultimate flow control. Replaceable Boroflo PTFE Screw-Thread Keys create a chemically-resistant, leakproof system which requires no lubrication whatsoever. Each Borosil® Burette is calibrated by automatic laser calibration. Certificates can be downloaded by visiting Borosil.com, or by downloading the Borosil app. Avail. from 10mL to 100mL. Burettes comply with ISO-385. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.