Learn More
Thermo Scientific BURGENER MIRAMIST 0.4-2.0ML/MI
Catalog No. NC0235213 Shop All Thermo Scientific Products
Encompass
Burgener, MiraMist High Solids Nebulizer (0.4 - 2.0ml/min uptake rate)
This item is not returnable. View return policy
Catalog No. NC0235213 Supplier Thermo Scientific Supplier No. 4600356
Burgener, MiraMist High Solids Nebulizer (0.4 - 2.0ml/min uptake rate)